Learn More
Biomatik Corporation Human P-Selectin Glycoprotein Ligand 1 (PSGL1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU09039
The Human P-Selectin Glycoprotein Ligand 1 (PSGL1) ELISA Kit allows for quantitative determination of P-Selectin Glycoprotein Ligand 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.254 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14242. Cross Activity: No significant cross-reactivity or interference between P-Selectin Glycoprotein Ligand 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4328