Learn More
Biomatik Corporation Human Mitofusin 2 (MFN2) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU09220
The Human Mitofusin 2 (MFN2) ELISA Kit allows for quantitative determination of Mitofusin 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O95140. Cross Activity: No significant cross-reactivity or interference between Mitofusin 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4370