Learn More
Biomatik Corporation Human Nucleotide Binding Oligomerization Domain Containing Protein 2 (NOD2) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU09293
The Human Nucleotide Binding Oligomerization Domain Containing Protein 2 (NOD2) ELISA Kit allows for quantitative determination of Nucleotide Binding Oligomerization Domain Containing Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9HC29. Cross Activity: No significant cross-reactivity or interference between Nucleotide Binding Oligomerization Domain Containing Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4381