Learn More
Biomatik Corporation Human Meteorin Like Protein (METRNL) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU09365
The Human Meteorin Like Protein (METRNL) ELISA Kit allows for quantitative determination of Meteorin Like Protein in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.64 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q641Q3. Cross Activity: No significant cross-reactivity or interference between Meteorin Like Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4398