Learn More
Biomatik Corporation Rat Angiopoietin Like Protein 8 (ANGPTL8) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU09387
The Rat Angiopoietin Like Protein 8 (ANGPTL8) ELISA Kit allows for quantitative determination of Angiopoietin Like Protein 8 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: D3ZMI0. Cross Activity: No significant cross-reactivity or interference between Angiopoietin Like Protein 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4400