Learn More
Biomatik Corporation Human Annexin A1 (ANXA1) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09781
Encompass_Preferred
The Human Annexin A1 (ANXA1) ELISA Kit (CLIA) allows for quantitative determination of Annexin A1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.27-200 ng/mL. Sensitivity: 0.12 ng/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P04083. Cross Activity: No significant cross-reactivity or interference between Annexin A1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4430