Learn More
Biomatik Corporation Rat Butyrylcholinesterase (BCHE) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU10079
The Rat Butyrylcholinesterase (BCHE) ELISA Kit allows for quantitative determination of Butyrylcholinesterase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.63 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9JKC1. Cross Activity: No significant cross-reactivity or interference between Butyrylcholinesterase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4460