Learn More
Biomatik Corporation Mouse Hepcidin (Hepc) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU10127
The Mouse Hepcidin (Hepc) ELISA Kit allows for quantitative determination of Hepcidin in Serum, Plasma, Urine and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 25.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9EQ21. Cross Activity: No significant cross-reactivity or interference between Hepcidin and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.