Learn More
Biomatik Corporation High Sensitive Human Myelin Basic Protein (MBP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU10305
The High Sensitive Human Myelin Basic Protein (MBP) ELISA Kit allows for quantitative determination of Myelin Basic Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02686. Cross Activity: No significant cross-reactivity or interference between Myelin Basic Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4515