Learn More
Biomatik Corporation High Sensitive Human VGF Nerve Growth Factor Inducible (VGF) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU10325
The High Sensitive Human VGF Nerve Growth Factor Inducible (VGF) ELISA Kit allows for quantitative determination of VGF Nerve Growth Factor Inducible in Serum, Plasma, cerebrospinal fluid and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O15240. Cross Activity: No significant cross-reactivity or interference between VGF Nerve Growth Factor Inducible and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4520