Learn More
Biomatik Corporation Instant Rat Procollagen I N-Terminal Propeptide (PINP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU10371
The Instant Rat Procollagen I N-Terminal Propeptide (PINP) ELISA Kit allows for quantitative determination of Procollagen I N-Terminal Propeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 246.9-20000 pg/mL. Sensitivity: 92.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 1 hours, 10minutes. UniProt ID: P02454. Cross Activity: No significant cross-reactivity or interference between Procollagen I N-Terminal Propeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4521