Learn More
Biomatik Corporation Mouse Lactoferrin (LTF) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05539
The Mouse Lactoferrin (LTF) ELISA Kit allows for quantitative determination of Lactoferrin in Serum, Plasma, Breast milk and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.247 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08071. Cross Activity: No significant cross-reactivity or interference between Lactoferrin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3097