Learn More
Biomatik Corporation Human Nad-Dependent Deacetylase Sirtuin-1 (Sirt1/Sir2L1) Elisa Kit
Supplier:  Biomatik Corporation EKC34727
Encompass_Preferred
Human Nad-Dependent Deacetylase Sirtuin-1 (Sirt1/Sir2L1) Elisa Kit for detecting Sirtuin (Silent Mating Type Information Regulation 2 Homolog) 1 (S. Cerevisiae) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q96EB6; SIRT1. Target Synonyms: 75SirT1; hSIR2; hSIRT1; HST2; HST2; S. cerevisiae; homolog of; NAD dependent deacetylase sirtuin 1; NAD dependent protein deacetylase sirtuin 1; NAD-dependent deacetylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; Regulatory protein SIR2 homolog 1; SIR1_HUMAN; SIR2; SIR2 like 1; SIR2 like protein 1; SIR2; S.cerevisiae; homolog-like 1; SIR2-like protein 1; SIR2ALPHA; SIR2L1; Sirt1; SirtT1 75 kDa fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7487
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.