Learn More
Biomatik Corporation Human Collagen Type I (COL1) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11911
Encompass_Preferred
Human Collagen Type I (COL1) Anti-antigen ELISA Kit for detecting COL1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.21ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P02452. Gene ID: 1277. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7026
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.