Learn More
Biomatik Corporation Mouse GLUT1 (Glucose Transporter 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58314
Encompass_Preferred
Mouse GLUT1 (Glucose Transporter 1) ELISA Kit for detecting GLUT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17809. Target Synonyms: GLUT1; Glucose Transporter 1; SLC2A1; Solute carrier family 2; facilitated glucose transporter member 1; Glucose transporter type 1; erythrocyte; brain; GLUT-1; HepG2 glucose transporter; GLUT1; PED; DYT9; GLUT; DYT17; DYT18; EIG12; GLUT1DS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8094
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.