Learn More
Fisherbrand™ Elite™ Multichannel Pipetters
SCI_ED
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort.
 View more versions of this product
Catalog No. S98642A
Add to Cart
Description
Description
  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Elite™
5 to 50 μL
12
Pipette
3 years
Product Suggestions
Product Suggestions