Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elk-1 Mouse anti-Human, Clone: PCRP-ELK1-1D9, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Elk-1
Clone PCRP-ELK1-1D9
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Western Blot, Immunofluorescence, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121766
View Documents
Novus Biologicals
NBP30714620UG
20 μg
Each for $138.37
Add to cart
 
NB121765
View Documents
Novus Biologicals
NBP30714610UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Elk-1 Monoclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western Blot, Immunofluorescence, Protein Array
Specifications

Specifications

Elk-1
0.2 mg/ml
Western Blot, Immunofluorescence, Peptide Array
Purified
RUO
Human
ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
Recombinant full-length human Elk-1 protein
Monoclonal
Protein A or G purified
PCRP-ELK1-1D9
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific
10 mM PBS with 0.05% BSA
2002
IgG2b
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit