Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELK3 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI1H3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270643APCCY7

Catalog No. N270643APC7


Add to cart

Description

Description

ELK3 Monoclonal antibody specifically detects ELK3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ELK3
OTI1H3
Flow Cytometry
Affinity Purified, Conjugated
ELK3
Full length recombinant protein of human ELK3 (NP_005221) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ELK3, ETS-domain protein (SRF accessory protein 2), ERPETS domain-containing protein Elk-3, ETS-domain protein, ETS-related protein ERP, NETSRF accessory protein 2, SAP-2, SAP2ETS-related protein NET, Serum response factor accessory protein 2
Mouse
IgG1
0.1 mL
Primary
2004.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit