Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELK3 Mouse anti-Human, Mouse, Rat, APC, Clone: OTI1H3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270643APC
Description
ELK3 Monoclonal specifically detects ELK3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
ELK3 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
ELK3 | |
Full length recombinant protein of human ELK3 (NP_005221) produced in HEK293T cells. | |
0.1 mL | |
2004.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1H3 | |
Flow Cytometry | |
ELK3, ETS-domain protein (SRF accessory protein 2), ERPETS domain-containing protein Elk-3, ETS-domain protein, ETS-related protein ERP, NETSRF accessory protein 2, SAP-2, SAP2ETS-related protein NET, Serum response factor accessory protein 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title