Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELK3 Mouse anti-Canine, Human, Clone: OTI3D9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525694

Catalog No. PIMA525694


Add to cart

Description

Description

The protein encoded by this gene is a member of the ETS-domain transcription factor family and the ternary complex factor (TCF) subfamily. Proteins in this subfamily regulate transcription when recruited by serum response factor to bind to serum response elements. This protein is activated by signal-induced phosphorylation; studies in rodents suggest that it is a transcriptional inhibitor in the absence of Ras, but activates transcription when Ras is present.
Specifications

Specifications

ELK3
Monoclonal
0.78 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P41970
ERP, NET, SAP2, ETS domain-containing protein Elk-3, ETS-domain protein, ETS-related protein ERP, ETS-related protein NET, SAP-2, SRF accessory protein 2, serum response factor accessory protein 2
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2004, 482613
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3D9
Unconjugated
ELK3
Liquid
ELK3
Full length human recombit protein of ELK3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit