Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELK4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen ELK4
Immunogen Synthetic peptides corresponding to ELK4 (ELK4, ETS-domain protein (SRF accessory protein 1)) The peptide sequence was selected from the C terminal of ELK4. Peptide sequence ASLTPAFFSQVACSLFMVSPLLSFICPFKQIQNLYTQVCFLLLRFVLERL.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1692920 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16912920UL
20ul Each for $149.00
Add to cart
 
NBP169129 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169129
100 ul Each for $329.00
Add to cart
 
Description

Description

ELK4 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ELK4
Rabbit
Polyclonal
Unconjugated
2005
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to ELK4 (ELK4, ETS-domain protein (SRF accessory protein 1)) The peptide sequence was selected from the C terminal of ELK4. Peptide sequence ASLTPAFFSQVACSLFMVSPLLSFICPFKQIQNLYTQVCFLLLRFVLERL.
Primary
IgG
ELK4, ETS-domain protein (SRF accessory protein 1), ETS-domain protein, SAP-1, SAP1ETS domain-containing protein Elk-4, Serum response factor accessory protein 1, SRF accessory protein 1
PBS & 2% Sucrose. with No Preservative
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit