Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELL Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30394420UL

 View more versions of this product

Catalog No. NB094901


Add to cart

Description

Description

ELL Polyclonal antibody specifically detects ELL in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

ELL
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot
PBS with 50% glycerol, pH7.3.
C19orf17, chromosome 19 open reading frame 17, DKFZp434I1916, Eleven-nineteen lysine-rich leukemia protein, ELL gene (11-19 lysine-rich leukemia gene), ELL1, elongation factor RNA polymerase II, MEN, RNA polymerase II elongation factor ELL
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human ELL (NP_006523.1).
Affinity purified
RUO
Primary
8178
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit