Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen ELL2
Immunogen Synthetic peptides corresponding to ELL2 (elongation factor, RNA polymerase II, 2) The peptide sequence was selected from the C terminal of ELL2. Peptide sequence PGSKEYQNVHEEVLQEYQKIKQSSPNYHEEKYRCEYLHNKLAHIKRLIGE.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16922020 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16922020UL
20ul Each for $149.00
Add to cart
 
NBP169220 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169220
100 ul Each for $329.00
Add to cart
 
Description

Description

ELL2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ELL2
Rabbit
Polyclonal
Unconjugated
22936
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to ELL2 (elongation factor, RNA polymerase II, 2) The peptide sequence was selected from the C terminal of ELL2. Peptide sequence PGSKEYQNVHEEVLQEYQKIKQSSPNYHEEKYRCEYLHNKLAHIKRLIGE.
Primary
IgG
ELL-related RNA polymerase II, elongation factor, elongation factor, RNA polymerase II, 2, RNA polymerase II elongation factor ELL2
PBS & 2% Sucrose. with No Preservative
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit