Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elma™ Clean 260 Dip & Splash Ultrasonic Cleaning Solution

Safely cleans sensitive metals, plastics and glass in an ultrasonic cleaner or labware washer.

Manufacturer:  Elma™ 800 0029

 View more versions of this product

Catalog No. 08-000-150


Add to cart

Alert:

Irritation to eyes.


Description

Description

  • Neutral pH concentrate
  • Leaves no residue after rinsing with DI water
  • Dilute to 2% for use in an ultrasonic cleaner or to 1% for a labware washer
  • Dilute with either tap water or deionized water
  • Biodegradable
  • Available in 10 and 25L containers
Applications
  • Removes coolant, oil, dust, fingerprints, marking and label residues and soap deposits
  • Cleans aluminum surfaces prior to anodizing or coating
  • Cleans laser machined parts, laboratory sieves, sensitive metal, plastic, glass or ceramic surface
Specifications

Specifications

Safely cleans sensitive metals, plastics and glass in an ultrasonic cleaner or labware washer
Keep between 43° and 85°F, shelf life 2 years
Liquid
25L
Neutral pH
Ultrasonic Cleaning Product
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit