Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELMOD1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548943

Catalog No. PA548943

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Acts as a GTPase-activating protein (GAP) toward guanine nucleotide exchange factors like ARL2, ARL3, ARF1 and ARF6, but not for GTPases outside the Arf family.
Specifications

Specifications

ELMOD1
Polyclonal
PBS with 0.09% sodium azide
Q8N336, Q3V1U8
ELMO domain-containing protein 1, ELMOD1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
270162, 55531
Western Blot
Unconjugated
ELMOD1
Liquid
ELMOD1
KLH conjugated synthetic peptide between 106-135 amino acids from the Central region of human ELMOD1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit