Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELMOD2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ELMOD2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096389
View Documents
Novus Biologicals
NBP305278100UL
100 μg Each for $320.71
Add to cart
 
NB096390
View Documents
Novus Biologicals
NBP30527820UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ELMOD2 Polyclonal antibody specifically detects ELMOD2 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

ELMOD2
Unconjugated
Rabbit
Apoptosis
PBS with 50% glycerol, pH7.3.
255520
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Mouse
ELMO domain containing 2, ELMO domain-containing protein 2,9830169G11Rik, ELMO/CED-12 domain containing 2, FLJ35768, FLJ38038, MGC10084
Recombinant protein of mouse ELMOD2
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit