Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30389520UL

 View more versions of this product

Catalog No. NB094154


Add to cart

Description

Description

ELOVL4 Polyclonal antibody specifically detects ELOVL4 in Human samples. It is validated for Western Blot
Specifications

Specifications

ELOVL4
Unconjugated
Purified
Rabbit
IgG
20 μL
Lipid and Metabolism, Vision
Primary
6785
Western Blot
PBS with 50% glycerol, pH7.3.
ADMD, cancer/testis antigen 118, CT118, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4, elongation of very long chain fatty acids protein 4, FLJ17667, FLJ92876,3-keto acyl-CoA synthase ELOVL4, Stargardt disease 3 (autosomal dominant), STGD2, STGD3
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ELOVL4 (NP_073563.1).
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit