Learn More
ELOVL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 211601AP
Catalog No. 50-173-3289
Add to cart
Description
Description
Lipogenesis is a key event in the energy storage system and is controlled by the transcription factor sterol regulatory element-binding protein (SREBP)-1. Elongation of very long chain fatty acids protein 6 (ELOVL6) is a member of fatty acyl-CoA elongase gene family that converts palmitic to stearic acid and it has been shown to be a target of SREBP-1, playing an important role in de novo synthesis of long-chain saturated and monosaturated fatty acids in conjunction with fatty acid synthase and stearoyl-CoA desaturase ELOVL6 was predicted to be important for tissue fatty acid composition. Recent studies suggest that inhibition of this elongase could be a new therapeutic approach for ameliorating insulin resistance, diabetes and cardiovascular risks, even in the presence of a continuing state of obesity.
Specifications
Specifications
ELOVL6
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q920L5, Q920L6, Q9H5J4
ELOVL6, FACE, FAE, Fatty acid elongase 2, Fatty acyl CoA elongase, hELO2, LCE, Long chain fatty acyl elongase
Rabbit
IgG
150 μL
-20°C
Primary
170439, 171402, 79071
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ELOVL6
Liquid
Elovl6
ELOVL6 Fusion Protein Ag14163
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat