Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550352

Catalog No. PA550352


Add to cart

Description

Description

The antibody detects endogenous levels of total ELP2 protein.

ELP2 regulates the ligand-dependent activation of STAT3. The protein acts as subunit of the RNA polymerase II elongator complex, which is a histone acetyltransferase component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4.
Specifications

Specifications

ELP2
Polyclonal
Unconjugated
ELP2
Liquid
ELP2
Fusion protein corresponding to residues near the C terminal of human elongator acetyltransferase complex subunit 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.9 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q6IA86
STATIP1
Rabbit
IgG
100 μL
-20°C
Primary
55250
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit