Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elp3 Mouse anti-Human, DyLight 488, Clone: 11811, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243694G

Catalog No. NBP243694G


Add to cart

Description

Description

Elp3 Monoclonal antibody specifically detects Elp3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Elp3
11811
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
ELP3
Recombinant protein encompassing a sequence within the center region of human ELP3. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.3.1.48, elongation protein 3 homolog (S. cerevisiae), elongator complex protein 3, hELP3, KAT9FLJ10422
Mouse
IgG1
0.1 ml
Primary
55140
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit