Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELTD1/ETL Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $378.00
Specifications
Antigen | ELTD1/ETL |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ELTD1/ETL Polyclonal specifically detects ELTD1/ETL in Human samples. It is validated for Western Blot.Specifications
ELTD1/ETL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
EGF, latrophilin and seven transmembrane domain containing 1, EGF, latrophilin and seven transmembrane domain-containing protein 1, ETL protein, ETLEGF-TM7-latrophilin-related protein, KPG_003 | |
ADGRL4 | |
IgG | |
Affinity Purified |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
GPCR, Neuroscience | |
PBS with 0.02% Sodium Azide | |
64123 | |
Reacts with residues 7-19 [RSSSNQDRFITND] of the human ETL protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title