Learn More
Invitrogen™ ELTD1 Monoclonal Antibody (CL4164)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524705
Description
Immunogen sequence: ENANCTNTEGS YYCMCVPGFR SSSNQDRFIT NDGTVCIENV NANCHLDNVC IAANINKTLT KIRSIKEPVA LLQEVYRNSV TDLSPTDIIT YIEILAESSS LLGYKNNTIS Binds to an epitope located within the peptide sequence CVPGFRSSSN as determined by overlapping synthetic peptides. Highest antigen sequence identity to the following orthologs - mouse 53%, rat 55%.
ETL is a novel seven-transmembrane receptor that belongs to the secretin family of G-protein-coupled peptide hormone receptors and the EGF-TM7 subfamily of receptors. It is the first seven-transmembrane receptor containing EGF-like repeats that is developmentally regulated in the heart.
Specifications
ELTD1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBW9 | |
ADGRL4 | |
Recombinant Human ELTD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
CL4164 | |
Unconjugated | |
ADGRL4 | |
1110033N21Rik; ADGRL4; Adhesion G protein-coupled receptor L4; EGF, latrophilin and seven transmembrane domain containing 1; EGF, latrophilin and seven transmembrane domain-containing protein 1; EGF, latrophilin seven transmembrane domain containing 1; EGF, latrophilin seven transmembrane domain-containing protein 1; EGF,latrophilin and seven transmembrane domain-containing protein 1; EGF-TM7 receptor; EGF-TM7-latrophilin-related protein; Eltd1; ETL; ETL protein; ETL1; hCG_14667; KPG_003; UNQ202/PRO228 | |
Mouse | |
Protein A | |
RUO | |
64123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.