Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eltoprazine hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
98206-09-8
C12H17ClN2O2
256.73
JFSOSUNPIXJCIX-UHFFFAOYSA-N
hydrogen eltoprazine chloride
3025067
1-(2,3-dihydro-1,4-benzodioxin-5-yl)piperazine;hydrochloride
C1CN(CCN1)C2=C3C(=CC=C2)OCCO3.Cl
This item is not returnable. View return policy
98206-09-8
C12H17ClN2O2
256.73
JFSOSUNPIXJCIX-UHFFFAOYSA-N
hydrogen eltoprazine chloride
3025067
1-(2,3-dihydro-1,4-benzodioxin-5-yl)piperazine;hydrochloride
C1CN(CCN1)C2=C3C(=CC=C2)OCCO3.Cl

5-HT1 receptor agonist/partial agonist

5-HT 1 and 5-HT 2C receptor partial agonist (K i values are 40, 52 and 81 nM for 5-HT 1A 5-HT 1B and 5-HT 2C receptors respectively). Reduces 5-HIAA levels in the striatum and exhibits antiaggressive behavior in vivo.
Quantity 10mg
Assay Percent Range >98%
Formula Weight 256.73
Chemical Name or Material Eltoprazine hydrochloride

Recommended Storage : Store at Room Temperature