Learn More
Eluent Solution, for Ion Chromatography, 2.8mM NaHCO-Na2CO3, Ricca Chemical
Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 28131
Packaging Poly natural
Quantity 4L
 View more versions of this product
Catalog No. 2813-1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
7732-18-5,144-55-8,497-19-8
100.0,0.02,0.02
Eluent Solution, 2.8mM NaHCO
98.0,0.02,0.02
Liquid
Specifications
7732-18-5,144-55-8,497-19-8
100.0,0.02,0.02
Poly natural
4L
98.0,0.02,0.02
Liquid
Clear colorless
Eluent Solution, 2.8mM NaHCO