Learn More
FUJIFILM Wako ELX808 Microplate Reader and 21 CFR Part 11, ERES Compliant Set
Set containing the Biotek ELx808IU microplate reader, laptop computer and Toximaster™ QC8 software
Manufacturer:  FUJIFILM Wako BIOT35931
Catalog No. 12-333-049
Add to cart
Description
Description
  • 96-well configuration
  • Ideal for high throughput of samples
  • Superior 4-Zone™ incubation
  • Fast kinetics, endpoint and linear well scanning
  • Optical design eliminates crosstalk
  • Extensive endotoxin analysis with Toximaster™ QC8 software
Specifications
Specifications
Absorbance Spectrophotometer
Biotek
Option of Gen5Secure of Toximaster™ QC8
Plastic, Endotoxin-free, 96-wells
340 to 900 nm
Available
As fast as 6 sec.
Biotek ELx808IU
±2.0% from 0.000 to 2.500 OD at 405 nm
High power LED
Single, dual or kinetic
No
Variable up to 50 °C
96 wells: 12 sec. (single wavelength), 20 sec. (dual wavelength), or 6 sec. (kinetic single wavelength)
Biotek ELx808IU microplate reader, Laptop computer, Toximaster™ QC8 software
ERES compliant