Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat EMAP-II Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1910100
This item is not returnable.
View return policy
Description
EMAP-II/AIMP1 Monoclonal specifically detects EMAP-II/AIMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
EMAP-II/AIMP1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AIMP1, aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPII, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43, SCYE1 | |
E.coli-derived recombinant human EMAP-II Ser147-Lys312 Accession # Q12904 | |
100 μg | |
Primary | |
Detects human EMAP-II in direct ELISAs. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
959516 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q12904 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9255 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction