Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMAP-II/AIMP1 Mouse anti-Human, Mouse, Rat, Clone: 959516, R&D Systems™

Mouse Monoclonal Antibody

$113.85 - $339.00

Specifications

Antigen EMAP-II/AIMP1
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Format Purified
Clone 959516
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1910SP View Documents R&D Systems
MAB1910SP
25 ug Each for $113.85
Add to cart
 
MAB1910100 View Documents R&D Systems
MAB1910100
100 ug Each for $339.00
Add to cart
 
Description

Description

EMAP-II is a pro-inflammatory cytokine that is chemotactic for monocytes and granulocytes. It is a cleavage product of multisynthetase complex auxiliary component p43.
Specifications

Specifications

EMAP-II/AIMP1
Purified
Immunocytochemistry, Immunohistochemistry, Western Blot
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
959516
Unconjugated
IgG2b
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit