Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EMC6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564051
Description
Immunogen sequence: VVAKREGPPF ISEAAVRGNA AVLDYCRTS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
TMEM93 belongs to the TMEM93 family. It is a multi-pass membrane protein. The function of the TMEM93 protein remains unknown.
Specifications
EMC6 | |
Polyclonal | |
Unconjugated | |
EMC6 | |
EMC6; ER membrane protein complex subunit 6; TMEM93; Transmembrane protein 93 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BV81 | |
EMC6 | |
Recombinant protein corresponding to Human EMC6. Recombinant protein control fragment (Product #RP-103464). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction