Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMC7 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260710
Description
EMC7 Polyclonal specifically detects EMC7 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative).Specifications
EMC7 | |
Polyclonal | |
Western Blot 1:1000, Immunoprecipitation | |
C11orf3, C15orf24, chromosome 15 hypothetical ATG/GTP binding protein, chromosome 15 open reading frame 24, ER membrane protein complex subunit 7, HT022, ORF1-FL1 | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
EMC7 | |
between 1 and 50 | |
100 μL | |
56851 | |
Store at 4degreesC. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title