Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4NB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550212

Catalog No. PA550212


Add to cart

Description

Description

The antibody detects endogenous levels of total EMC8 protein.

Expressed in liver. pancreas. heart. lung. kidney. brain. skeletal muscle. and placenta. Expression levels are highest in pancreas and moderate in heart. skeletal muscle. and placenta.
Specifications

Specifications

COX4NB
Polyclonal
Unconjugated
EMC8
Liquid
EMC8
Fusion protein corresponding to a region derived from internal residues of human ER membrane protein complex subunit 8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O43402
C16orf2, C16orf4, COX4AL, COX4NB, FAM158B, NOC4
Rabbit
IgG
100 μL
-20°C
Primary
10328
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit