Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EMD 386088 hydrochloride

Potent 5-HT6 agonist

$164.00 - $647.50

Specifications

Assay Percent Range >99%
Formula Weight Observed MW: 283.2
Chemical Name or Material 5-Chloro-2-methyl-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole hydrochloride
Molecular Formula C14H15N2Cl.HCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-825-0 View Documents Tocris Bioscience™
2382/50
50mg Each for $647.50
Add to cart
 
23-821-0 View Documents Tocris Bioscience™
2382/10
10mg Each for $164.00
Add to cart
 
Description

Description

Potent 5-HT6 receptor agonist (EC50 = 1.0 nM) that displays selectivity over other 5-HT receptors (IC50 values are 7.4, 110, 180, 240, 450, 620, 660 and 3000 nM for 5-HT6, 5-HT1D, 5-HT1B, 5-HT2A, 5-HT2C, 5-HT4, 5-HT1A and 5-HT7 receptors respectively). Shows moderate affinity at 5-HT3 receptors (IC50 = 34 nM).
Specifications

Specifications

>99%
Observed MW: 283.2
5-Chloro-2-methyl-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole hydrochloride
C14H15N2Cl.HCl
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at RT

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit