Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ 4EBP1, HisTag™, Human, Recombinant Protein, S. frugiperda
Supplier: MilliporeSigma™ 51667850UG
Description
Full-length, recombinant, human 4EBP1 fused at the N-terminus to a His·Tag™ sequence and expressed in S. frugiperda insect cells. 4EBP1 negatively regulates translation and is inhibited by p70S6K phosphorylation. 4EBP1 is useful as a substrate in the study of p70S6K regulation and inhibitor screening.Specifications
Concentration on Label | |
SDS-PAGE | |
S. frugiperda | |
Following initial thaw, aliquot and freeze (−70°C). | |
His-Tag | |
Recombinant | |
≥75% |
In 300mM NaCl, 150mM Imidazole, 50mM Sodium Phosphate, 0.2mM DTT, 0.1mM PMSF, 25% Glycerol, pH 7.0. | |
50μg | |
4EBP1 | |
RUO | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction