Learn More
EMD Millipore Amicon™ Bioseparations Stirred Cells† Available on GSA/VA Contract for Federal Government customers only.
Concentrate, desalt, and purify aqueous biological solutions
Manufacturer:  EMD Millipore 5121
 View more versions of this product
Catalog No. 5121
  Check Availability
Add to cart
Requires : Magnetic stirrer required for operation.
Specifications Description & Specifications
Specifications
Yes
1 to 10mL
0.05 to 10g%
2.5cm
4.1cm2
3 in.
2.38 in.
Nylon cap and tube fitting assembly, Polysulfone cylinder and membrane holder, Acetal and Polysulfone stirring assembly, Silicone Rubber O-rings, Polyethelene pressure tube, Tygon tubing filtrate tube, Anodized Aluminum or Nylon retaining stand
200μL
6cm
0.03 to 0.3 oz.
0.03 to 0.3 oz., 1 to 10mL
0.9 in.
2.38 x 2.38 x 3 in. (6 x 6 x 7.7cm)
Protein purification, protein concentration, protein desalting
7.7cm
6cm
8010
2.38 in.
Clear pressurized vessel with integral magnetic stirring bar, nylon cap and tube fitting assembly, polysulfone body and membrane holder, acetal/polysulfone stirrer, silicone O-rings and black aluminum stand, connections for 0.25 in. dia. pressure tubing