Learn More
EMD Millipore Amicon™ Bioseparations Stirred Cells†
Concentrate, desalt, and purify aqueous biological solutions
Manufacturer:  EMD Millipore 5122
 View more versions of this product
Catalog No. 5122
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Requires : Magnetic stirrer required for operation.
Specifications Description & Specifications
Specifications
Yes
2.5 to 50mL
0.05 to 10g%
4.45cm
13.4cm2
3.8 in.
2.7 in.
Nylon cap and tube fitting assembly, Polysulfone cylinder and membrane holder, Acetal and Polysulfone stirring assembly, Silicone Rubber O-rings, Polyethelene pressure tube, Tygon tubing filtrate tube, Anodized Aluminum or Nylon retaining stand
500μL
7cm
0.08 to 1.6 oz.
0.08 to 1.6 oz., 2.5 to 50mL
1.7 in.
2.7 x 2.7 x 3.8 in. (7 x 7 x 9.8cm)
Protein purification, protein concentration, protein desalting
9.8cm
7cm
8050
2.7 in.
Clear pressurized vessel with integral magnetic stirring bar, nylon cap and tube fitting assembly, polysulfone body and membrane holder, acetal/polysulfone stirrer, silicone O-rings and black aluminum stand, connections for 0.25 in. dia. pressure tubing