Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 3 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
Manufacturer:  MilliporeSigma™ ACS500302
 View more versions of this product
Catalog No. ACS500302
Add to cart
Description
Description
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.
Specifications
Specifications
Yes
3 KDa
Regenerated Cellulose (RC)
12 cm
2Pack
15 mL
Non-sterile
Centrifugal Filters
2 Amicon Ultra NMWCO 3 kDa filters, 2 Exchange devices, and 2 Collection tubes with cap