EMD Millipore™ Atrial Natriuretic Factor 1-28, Human Neuropeptide, Calbiochem

Manufacturer: EMD Millipore 0523030005MG

 View all options for this product

Catalog No. 05-230-30005M

  Check Availability
Add to Cart


    Purity ≥97%
    Purification Method HPLC
    Quantity 0.5mg
    Format White to off-white lyophilized solid
    Formulation Supplied as a trifluoroacetate salt.
    Regulatory Status RUO
    Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 month at −20°C.
    Product Type Atrial Natriuretic Factor 1-28
    CAS 91917-63-4

    Increases the rate of renal excretion. Higher levels are reported in congestive heart failure. Inhibits MAP kinase through the natriuretic peptide clearance receptor.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit