Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Aurora A Kinase, HisTag, Human, Recombinant Protein, S. frugiperda
Manufacturer: MilliporeSigma 48141310UG
Description
Full-length, recombinant, human Aurora A kinase (amino acids 2-403) (GenBank Accession: NM_003600) fused at the N-terminus to a His·Tag sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. Aurora A plays an important role in chromosome segregation and cell division.Specifications
Concentration on Label | |
In 100mM NaCl, 25mM Tris-HCl, 3mM DTT, 50% glycerol, 0.05% Tween™-20 Detergent, pH 7.5. | |
≥90% | |
S. frugiperda | |
Following initial thaw, aliquot and freeze (−70°C). | |
His·Tag™ | |
Yes |
Liquid | |
SDS-PAGE | |
10μg | |
Aurora A Kinase | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title