Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Automation Compatible Filters

Trouble-free performance on a variety of automated systems

Manufacturer:  MilliporeSigma™ SLCRDZ5NK

 View more versions of this product

Catalog No. SLCRDZ5NK


Add to cart

Description

Description

Use with Zymark™ BenchMate™, TPW™ II, and MultiDose™ workstations, as well as Zymate™ stations that use filtration.

  • Pressure-resistant, high-density polyethylene housing prevents instrument stoppage
  • Some membranes also include a borosilicate glass fiber prefilter to process solutions containing high particle levels
  • Nonsterile

  • Hydrophilic Durapore™ PVDF membrane: For lowest protein binding sterilization/clarification of aqueous and mild organic solutions
  • Hydrophilic PTFE membrane: Low protein binding, LCR membrane for processing and clarifying aqueous and organic solutions
  • Nylon membrane: Broad chemical compatibility for processing and clarifying aqueous and organic solutions
  • Glass Fiber B Prefilter: For processing and clarifying aqueous and organic solutions containing high particle levels

Specifications

Specifications

Syringe Filter
Polytetrafluoroethylene (PTFE)
<200μ (for units with membrane and prefilter)
0.45μm
1000/pk.
25mm
100psi
<>100mL
3.9cm sq.
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit