Learn More
MilliporeSigma™ Calbiochem™ Benzalkonium Chloride
Cationic surface-active agent and germicide.
Supplier:  MilliporeSigma™ 198901250ML
Catalog No. 19-890-1250ML
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Its effects are potentiated in the presence of di-n-butyl esters, such as diisobutyladipate. M.W. ∽360.
Specifications
Specifications
15-30°C
250 mL