EMD Millipore Calbiochem™ Biotin-X-NHS Available on GSA/VA Contract for Federal Government customers only.

Used to biotinylate amino acids, peptides or proteins by reacting with primary amines under mild conditions

Manufacturer: EMD Millipore 20318825MG

 View more versions of this product

Catalog No. 20-318-825MG

  Check Availability
Add to Cart

    Specifications

    Quantity 25mg
    Description Biotin-X-NHS

    • Inserts a 6-atom spacer between biotin and the target ligand, thereby alleviating steric hindrance
    • Useful for red blood cell survival studies
    • Form: White solid
    • Molar Mass: 454.5
    • Purity: ≥90% by NMR
    • Soluble in DMSO

    Storage:

  • Store at 2° to 8°C. Protect from moisture.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit